Order Entry
Ireland
ContactUsLinkComponent
Safety spectacles, Horizon Flip Up
Safety spectacles, Horizon Flip Up
Catalog # DALL3024042-STD
Supplier:  HONEYWELL SAFETY
undefined
Safety spectacles, Horizon Flip Up
Catalog # DALL3024042-STD
Supplier:  HONEYWELL SAFETY
Specifications
  • Type:
    Abrasive resistant, shade lens colour IR 5
  • Pk:
    1
Specifications
About this item
The flip up dual lens safety glasses feature a clear impact resistant PC lens and a flip up shaded lens (IR 5).
  • Ajustable temple wires and flip-up window
  • Includes an integrated and superior sub-orbital protection and moulded sideshields
  • Abrasive resistant
  • Shade IR 5
Ideal for welders and industrial workers who require a shaded lens while working with high intensity light sources and a traditional clear impact-resistant polycarbonate lens when their task is complete.
Certifications: ANSI Z87.1-2003