
4-Cyano-4-phenylcyclohexanone 97%
Catalog # L05281.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:25115-74-6
:
Formula:C₁₃H₁₃NO
:199.24 g/mol
Melting Pt:112…118 °C
EINECS:246-631-7
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD00044816