Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Caffeine 1000 µg/ml in methanol, Cerilliant
Organic reference standard, Caffeine 1000 µg/ml in methanol, Cerilliant
  PROOCERC-051
 :  
Organic reference standard, Caffeine 1000 µg/ml in methanol, Cerilliant
  PROOCERC-051
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    58-08-2
  • Description:
    Caffeine 1,000 µg/ml in methanol
  • Formula:
    C₈H₁₀N₄O₂
  • Description:
    Caffeine