(4-Nitrophenyl)-α-D-glucopyranoside ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3767-28-0
:
Formula:C₁₂H₁₅NO₈
MDL Number:MFCD00064088
:301.25 g/mol
Melting Pt:206…207 °C
EINECS:223-189-3
Related Information
Frequently Bought Together