Order Entry
Ireland
ContactUsLinkComponent
Calcium chloride cylinders
Calcium chloride cylinders
Catalog # WITG5.624.104
Supplier:  witeg Labortechnik
undefined
Calcium chloride cylinders
Catalog # WITG5.624.104
Supplier:  witeg Labortechnik
Specifications
  • Pk:
    1
  • Ø ext.×H:
    75 mm x 500 mm
Specifications
About this item
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes