Organic reference standard, Benzo[k]fluoranthene 100 µg/ml in acetonitrile
Catalog # PROONE5021
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,5 mL
- CAS No.:207-08-9
- Description:Benzo[k]fluoranthene 100 µg/ml in acetonitrile
- Formula:C₂₀H₁₂
- Description:Benzo[k]fluoranthene