Alizarine yellow R 70% (dye content), Sigma-Aldrich®
Catalog # 195073-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance (Color):Brown to Very Dark Brown
- Appearance (Form) :Powder
- Infrared spectrum:Conforms to Structure
- Wavelength:380 - 390 nm
- Extinction Coefficient:≥16000
- Extinction Coefficient, c = 0.006G/L, in Methanol:≥7000
- Carbon:≥37.0 %
- Nitrogen:≥9.9 %
- TLC:Conforms to Structure
- Solubility (Color), c = 1%, 1N NH4OH:Red to Very Dark Red
Specification Test Results
About this item
:
Melting Pt:>300 °C (lit.)
:M-Clarity™ quality level = MQ200
EINECS:218-818-3
:GHS07
CAS Number:2243-76-7
Formula:C₁₃H₉N₃O₅
MDL Number:MFCD00007313