Organic reference standard, L(-)-Carnitine 10 µg/ml in methanol
Catalog # PROONE9231
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:541-15-1
- Description:L(-)-Carnitine 10 µg/ml in methanol
- Formula:C₇H₁₅NO₃
- Description:L(-)-Carnitine