Cumene hydroperoxide 80%, Technical Grade
:80-15-9
:
:C₉H₁₂O₂
:MFCD00002129
:152.20 g/mol
:100…102 °C (8 mmHg)
:–30 °C
:1,030
:56 °C (132 °F)
:201-254-7
:3109
:5.2,II
:GHS06
:GHS08
:GHS05
:GHS02
:GHS07
:Refrigerator
- Appearance (Colour)::Clear colorless to pale yellow
- Form:Liquid
- Assay (Iodometric Titration):≥75%
- Identification (FTIR):Conforms
- Refractive Index:1,5170-1,5250 @ 20°C
Frequently Bought Together