4-Methylumbelliferone
Catalog # MOLE39596112-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:176.17 g/mol
EINECS:201-986-7
CAS Number:90-33-5
Formula:C₁₀H₈O₃
MDL Number:MFCD00006866