
4-Methylumbelliferone 97%
:90-33-5
:
:C₁₀H₈O₃
:MFCD00006866
:176.17 g/mol
:186…191 °C
:Ambient
:201-986-7
:6.1,III
:GHS07
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:186 to 191 °C
- Water:≤2.5 % (K.F.)
- Sulfated ash:≤0.1 %
- HPLC:≥96.0 %
- Titration with NaOH:≥96.0 % (On dry substance)
Frequently Bought Together