5-Bromo-2'-deoxyuridine (broxuridine), Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
Synonyms: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU
CAS Number:59-14-3
:
Formula:C₉H₁₁BrN₂O₅
MDL Number:MFCD00006529
:307.10 g/mol
EINECS:200-415-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together