Order Entry
Ireland
ContactUsLinkComponent
Potassium perfluorobutane-1-sulphonate for synthesis
Potassium perfluorobutane-1-sulphonate for synthesis
Catalog # 8.18662.0050
Supplier:  Merck
CAS Number:  
Potassium perfluorobutane-1-sulphonate for synthesis
Catalog # 8.18662.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    51 g/l (25 °C)
  • Melting Point:
    264 - 266 °C
  • Molar Mass:
    338.19 g/mol
  • Bulk Density:
    1000 kg/m3
  • pH:
    5.5 - 6.5 (50 g/l, H2O)
  • Synonyms:
    Nonafluorobutane-1-sulfonic acid potassium salt
  • Hill Formula:
    C4F9KO3S
  • Chemical Formula:
    CF3(CF2)3SO3K

Specification Test Results

About this item

Synonyms: Nonafluoro-1-butanesulphonic acid potassium salt, Potassium Perfluoro-1-butanesulphonate, Potassium Nonafluoro-1-butanesulphonate, Perfluoro-1-butanesulphonic acid potassium salt, Potassium nonafluoro-1-butanesulphonate
 :
 :338.1899 g/mol
EINECS:249-616-3
CAS Number:29420-49-3
Formula:C₄F₉KO₃S
MDL Number:MFCD00007515
 :M-Clarity™ quality level = MQ200