Dimethyl sulphoxide-[D₆] (99.8% D)
ROTHAE60.2
:
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
- Environmentally Preferable:
: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6
:
:84.09 g/mol
:189 °C (5 mmHg)
:20,2 °C
:1,19 g/cm³ (20 °C)
:82 °C (closed cup)
:Ambient
:218-617-0
:2206-27-1
:D₃CS(O)CD₃
:MFCD00002090