Desiccators with PROTEFAN safety coating
Supplier: witeg Labortechnik
Certificates
About this item
Desiccators are made of borosilicate glass, with safety PROTEFAN coating as implosion and splinter protection.
Frequently Bought Together
Desiccators are made of borosilicate glass, with safety PROTEFAN coating as implosion and splinter protection.