Skin graft blades
Supplier: SWANN MORTON
Certificates
About this item
Stainless steel. The 158 mm skin graft blades are used in the surgical removal of a patch of healthy skin from one area of the patient's body, usually the buttocks or inner thigh, which is then transplanted to another.
- Sterile
- Fit the Braithwaite (9901), Cobbett (9902) and Watson (9903) Skin Graft handles
Skin grafts may be recommended for burns, specific surgeries that may require skin grafts for healing to occur and for areas of prior infection with extensive skin loss.
Frequently Bought Together