Funnels, with standard ground joint cone
Supplier: witeg Labortechnik
Certificates
About this item
Funnels, made of boroslicate glass, with standard ground joint cone, one side flat.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.
Frequently Bought Together