Acriflavine hydrochloride
Catalog # MOLEM84900728
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:578.36 g/mol
CAS Number:8063-24-9
:69235-50-3
Formula:C₂₇H₂₈Cl₄N₆
MDL Number:MFCD00069039