Order Entry
Ireland
ContactUsLinkComponent
Pepstatin A
Pepstatin A
Supplier:  ENZO LIFE SCIENCES
Certificates
Pepstatin A
Supplier:  ENZO LIFE SCIENCES

Specifications

  • CAS No.:
    26305-03-3
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00060740
  • Protein/peptide name:
    Pepstatin A
  • Molecular formula:
    C₃₄H₆₃N₅O₉

Specifications

Frequently Bought Together