Order Entry
Ireland
ContactUsLinkComponent
Naphthalene Black 10B (Naphthol Blue Black, Acid Black 1)
Irritant
Naphthalene Black 10B (Naphthol Blue Black, Acid Black 1)
Catalog # ROTH9590.1
Supplier:  Roth Carl
CAS Number:  
Irritant
Naphthalene Black 10B (Naphthol Blue Black, Acid Black 1)
Catalog # ROTH9590.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :616.49 g/mol
EINECS:213-903-1
 :GHS07
Storage Temperature:Ambient
CAS Number:1064-48-8
Formula:C₂₂H₁₄N₆Na₂O₉S₂
MDL Number:MFCD00004017