Ponceau S 75% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:6226-79-5
:
Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number:MFCD00003892
:760.56 g/mol
:M-Clarity™ quality level = MQ200
EINECS:228-319-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Wavelength C=0.015 G/L DI water:518 - 522 nm
- Extinction Coefficient:≥33000
- Carbon:25.9 - 43.2 %
- Nitrogen:5.4 - 9.2 %
Specification Test Results
Frequently Bought Together