Order Entry
Ireland
ContactUsLinkComponent
 
Human ribonuclease inhibitor (from placenta)
 
undefined
Human ribonuclease inhibitor (from placenta)

 

  • Source:
    Placenta
  • Species:
    Human
  • Storage conditions:
    Freezer
  • Protein synonyms:
    RNase inhibitor
  • Protein/peptide name:
    Ribonuclease inhibitor

 

Frequently Bought Together