Order Entry
Ireland
ContactUsLinkComponent
(-)-Di-p-toluoyl-L-tartaric acid for synthesis, Sigma-Aldrich®
(-)-Di-p-toluoyl-L-tartaric acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
(-)-Di-p-toluoyl-L-tartaric acid for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    165 - 171 °C
  • Molar Mass:
    386.36 g/mol
  • Bulk Density:
    500 kg/m3

Specification Test Results

Related Information

Frequently Bought Together