Order Entry
Ireland
ContactUsLinkComponent
 
N-Acetyl-β-D-glucosamine
Catalog # ROTH8993.1
Supplier:  Roth Carl
CAS Number:  
N-Acetyl-β-D-glucosamine
Catalog # ROTH8993.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :221.21 g/mol
Storage Temperature:Ambient
EINECS:231-368-2
CAS Number:7512-17-6
Formula:C₈H₁₅NO₆
MDL Number:MFCD00136044