N-Acetyl-β-D-glucosamine
Catalog # MOLE20293490-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:221.21 g/mol
Storage Temperature:Ambient
EINECS:231-368-2
CAS Number:7512-17-6
Formula:C₈H₁₅NO₆
MDL Number:MFCD00136044