

Glutaryl dichloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2873-74-7
:
Formula:C₅H₆Cl₂O₂
MDL Number:MFCD00000755
:169.0059 g/mol
Density:1,324 g/cm³
Flash Pt:106 °C
Storage Temperature:Ambient
EINECS:220-711-1
UN:2922
ADR:8,II
:GHS06
:GHS05
Specification Test Results
- Appearance:Clear colorless to yellow to red-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4720 to 1.4760 (20°C, 589 nm)
- GC:≥96.0 %
- Titration RCOCl:≥96.0 %
Specification Test Results
Frequently Bought Together