(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
:96-47-9
:
:C₅H₁₀O
:MFCD00005367
:86.13 g/mol
:78…80 °C (760 mmHg)
:–136 °C
:0,86 g/cm³
:–11 °C
:Ambient
:202-507-4
:2536
:3,II
:GHS02
:GHS05
:GHS07
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.05 %
- GC:≥99.0 %
- Stabilizer:150 to 400 ppm BHT
Frequently Bought Together