Order Entry
Ireland
ContactUsLinkComponent
Sodium gluconate 98-102%, crystals, high purity
Sodium gluconate 98-102%, crystals, high purity
Catalog # E754-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium gluconate 98-102%, crystals, high purity
Catalog # E754-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    98 - 102 %
  • Heavy Metals:
    <0.001 %
  • Lead:
    <0.001 %
  • pH (10%, Water):
    6.8 - 7.5

Specification Test Results

About this item

 :
Merck Index:12,08766
Melting Pt:170…175 °C
 :218.14 g/mol
Storage Temperature:Ambient
EINECS:208-407-7
CAS Number:527-07-1
Formula:HOH₂C(CH(OH))₄COONa
MDL Number:MFCD00064210