Recombinant Taq DNA Polymerase
Catalog # 28-9373-48
Supplier: Cytiva
CAS Number:
Specifications
- Pk:25.000 EU
- Description:Recombinant Taq DNA Polymerase, 25000 units
- Seq:04
Specifications
About this item
Highly purified thermostable DNA polymerase for PCR.
- Highly purified recombinant enzyme
- Excellent batch-to-batch reproducibility
- Each lot is tested to be free of nuclease contamination