Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)
Catalog # ROTHK055.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:550.09 g/mol
Storage Temperature:Freezer
EINECS:243-347-5
CAS Number:19817-92-6
Formula:C₉H₁₂N₂Na₃O₁₅P₃
MDL Number:MFCD00044310