Order Entry
Ireland
Orders LinkContactUsLinkComponent
Uridine, Ultrapure

Uridine, Ultrapure

Catalog #
Uridine, Ultrapure
Catalog #
Supplier:  
CAS Number:  58-96-8
Uridine, Ultrapure
Uridine, Ultrapure
CAS Number:  58-96-8
Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Melting Pt:166…167 °C
  • Storage Temperature:Ambient
  • EINECS:200-407-5
Specifications

Specification Test Results

  • Em (261nm, 0.15M Phosphate Buffer, pH 7.3):
    ≥10100