Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Head Protection
Safety Helmets, customised
Safety helmet with logo IC, Vulcan concept
Safety helmet with logo IC, Vulcan concept
Safety helmet with logo IC, Vulcan concept
Catalog # CENUS22WA-IC
Supplier:
CENTURION SAFETY PRODUCTS
Expand image
Safety helmet with logo IC, Vulcan concept
Catalog # CENUS22WA-IC
Supplier:
CENTURION SAFETY PRODUCTS
Specifications
Colour:
White
Description:
Safety helmet with, Concept, with logo, IC, Vulcan
Environmentally Preferable:
Pk:
1
Specifications
About this item
Product Details
Product Details