Hand cleansing pastes, Plulux
Catalog # PLUM0718
Supplier: Plum Safety
Specifications
- Description:Hand cleansing paste, Plulux, bag-in-box, 1400 ml
- Pk:1
Specifications
About this item
Powerful hand cleansing paste. Effectively but gently removes ingrained and sticky grime such as diesel and hydraulic oil, oil mixed with soot, metal and cement dust.
- Apply on wet hands
- For dispenser systems MP2000
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904