Tubing connectors, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Unit consists of two barbed PE connectors, which assemble tightly together with a male-female centre taper.
- Suitable for vacuum applications
- Temperature resistant up to 80 °C
- Good chemical reistance
Frequently Bought Together