Order Entry
Ireland
ContactUsLinkComponent
Contact plate and Petri dish reader, SP Bel-Art
Contact plate and Petri dish reader, SP Bel-Art
Catalog # 710-0079
undefined
Contact plate and Petri dish reader, SP Bel-Art
Catalog # 710-0079
Specifications
  • Description:
    Contact plate and petri dish reader
  • Pk:
    1
Specifications
About this item
Made of polypropylene with a glass-clear acrylic deck for placement of plates and Petri dishes, this reader allows safe examination of the bottom of plates without risking spills on the face and eyes.
  • Ideal for examining cell monolayer, cell clones and for counting viral plaques or bacterial colonies
  • Easy and safe reading of hemagglutination, hemagglutination inhibition, hemolysis and agglutination tests
  • Clear deck for resting plates
  • Large, adjustable mirror is perfect for viewing plate underside
  • Mirror magnification is 3X actual plate size