3,6-Dimethyl-1,4-dioxane-2,5-dione ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: DL-Lactide
CAS Number:95-96-5
:
Formula:C₆H₈O₄
MDL Number:MFCD00011685
Boiling Pt:138…142 °C (8 mmHg)
Molecular Weight:144.13 g/mol
Melting Pt:122…126 °C
Flash Pt:180 °C (356 °F)
EINECS:202-468-3
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together