Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
2-(4-Methoxyphenoxy)-3-nitropyridine
2-(4-Methoxyphenoxy)-3-nitropyridine
2-(4-Methoxyphenoxy)-3-nitropyridine
Print
2-(4-Methoxyphenoxy)-3-nitropyridine
Catalog #
2-(4-Methoxyphenoxy)-3-nitropyridine
Catalog #
Supplier:
CAS Number:
76893-48-6
Expand image
Return to Top
2-(4-Methoxyphenoxy)-3-nitropyridine
Add to cart
CAS Number:
76893-48-6
Return Policy
Product Details & Documents
CAS Number:
76893-48-6
Formula:
C₁₂H₁₀N₂O₄
MDL Number:
MFCD00829021
Molecular Weight:
246.2188 g/mol