Organic reference standard, 3-Bromophenyl phenyl ether (BDE No. 2) 50 µg/ml in n-nonane, CIL
PROOCIL-BDE-2-CS
:
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:7025-06-1
- Description:3-Bromophenyl phenyl ether (BDE No. 2) 50 µg/ml in n-nonane
- Formula:C₁₂H₉BrO
- Description:3-Bromophenyl phenyl ether (BDE No. 2)