Hexachloroiridic(IV) acid hydrate (38 - 42% Ir) ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110802-84-1
:
Formula:H₂Cl₆Ir·xH₂O
MDL Number:MFCD00149906
ADR:8,III
:406.93 g/mol (anhy)
EINECS:241-012-8
UN:3260
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together