Order Entry
Ireland
ContactUsLinkComponent
Pesticide reference standard, 1,4-Dichloronaphthalene 100 µg/ml in n-nonane, CIL
Pesticide reference standard, 1,4-Dichloronaphthalene 100 µg/ml in n-nonane, CIL
  PROOCIL-ECN-2621
 :  
Pesticide reference standard, 1,4-Dichloronaphthalene 100 µg/ml in n-nonane, CIL
  PROOCIL-ECN-2621
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    1825-31-6
  • Description:
    1,4-Dichloronaphthalene 100 µg/ml in n-nonane
  • Formula:
    C₁₀H₆Cl₂
  • Description:
    1,4-Dichloronaphthalene