Order Entry
Ireland
ContactUsLinkComponent
Goggles, uvex 9305
Goggles, uvex 9305
Catalog # UVEX9305.714
Supplier:  UVEX
undefined
Goggles, uvex 9305
Catalog # UVEX9305.714
Supplier:  UVEX
Specifications
  • Type:
    Anti-fog
  • Colour:
    Translucent grey
  • Environmentally Preferable:
  • Lenses:
    Clear cellulose acetate
  • Pk:
    1
Specifications
About this item
Classic safety goggles with acetate lens.
  • Effective ventilation system
  • Adjustable textile headband
  • Mechanical strength: F –Low energy impact protection (45 m/s)
Certifications: EN 166, EN 170