Order Entry
Ireland
ContactUsLinkComponent
 
Purpurin (1,2,4-Trihydroxy-9,10-anthraquinone)
Catalog # MOLEM65000587
Supplier:  Molekula
CAS Number:  
Purpurin (1,2,4-Trihydroxy-9,10-anthraquinone)
Catalog # MOLEM65000587
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :256.21 g/mol
EINECS:201-359-8
CAS Number:81-54-9
Formula:C₁₄H₈O₅
MDL Number:MFCD00001203