Order Entry
Ireland
ContactUsLinkComponent
Malonic acid-[D₄] (99.5% D) for NMR spectroscopy
Irritant
Malonic acid-[D₄] (99.5% D) for NMR spectroscopy
Catalog # 8.15049.0025
Supplier:  Merck
CAS Number:  
Irritant
Malonic acid-[D₄] (99.5% D) for NMR spectroscopy
Catalog # 8.15049.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :108.03 g/mol
Merck Index:13,05732
EINECS:212-385-4
 :GHS07
Storage Temperature:Ambient
CAS Number:813-56-9
Formula:C₃D₄O₄