



Triethylborane 1 M in THF
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:97-94-9
:
Formula:C₆H₁₅B
MDL Number:MFCD00009022
Density:0,865
Flash Pt:–17 °C (1 °F)
Storage Temperature:Ambient
EINECS:202-620-9
UN:2924
ADR:3,II
:98 g/mol
:GHS02
:GHS05
:GHS08
:GHS07
Related Information
Frequently Bought Together