



Triethylborane 1 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:97-94-9
:
Formula:C₆H₁₅B
MDL Number:MFCD00009022
Density:0,86 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
EINECS:202-620-9
UN:2924
ADR:3,II
:98 g/mol
:GHS02
:GHS05
:GHS08
:GHS07
Specification Test Results
- Appearance:Colorless to amber Solution
- Strength:0.95 to 1.30 M
- Water:4.2 to 6.0 %
- HPLC:≥95 %
- pH:4.0 to 6.0
- Specific optical rotation:165° to 178° (20°C, 589 nm) (1 % in water DS)
Specification Test Results
Related Information
Frequently Bought Together