Hydrogen chloride 2 M in diethyl ether
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
:36.46 g/mol
Density:0,747
Flash Pt:–40 °C (–40 °F)
Storage Temperature:Refrigerator
EINECS:231-595-7
UN:2924
ADR:3,I
:GHS02
:GHS06
:GHS05
:GHS07
Related Information
Frequently Bought Together