

Chloroform-[D₁] stabilised for NMR spectroscopy
Catalog # ROTHAE59.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:120.38 g/mol
Boiling Pt:60,8 °C (1013 hPa)
Melting Pt:–64,1 °C
Density:1,5 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:212-742-4
UN:1888
ADR:6.1,III
:Stabilised with silver
:GHS08
:GHS07
CAS Number:865-49-6
Formula:CDCl₃
MDL Number:MFCD00000827