Cesium iodide ≥99.9995% (metals basis), TraceSELECT™, Fluka™
Catalog # 16722-25G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Aluminium (Al):Max. 0.02 ppm
- Arsenic (As):Max. 0.05 ppm
- Barium (Ba):Max. 0.1 ppm
- Beryllium (Be):Max. 0.005 ppm
- Calcium (Ca):Max. 0.1 ppm
- Cadmium (Cd):Max. 0.02 ppm
- Cerium (Ce):Max. 0.01 ppm
- Cobalt (Co):Max. 0.005 ppm
- Chromium (Cr):Max. 0.01 ppm
- Copper (Cu):Max. 0.005 ppm
- Dysprosium (Dy):Max. 0.01 ppm
- Erbium (Er):Max. 0.01 ppm
- Europium (Eu):Max. 0.01 ppm
- Iron (Fe):Max. 0.05 ppm
- Gallium (Ga):Max. 0.005 ppm
- Gadolinium (Gd):Max. 0.01 ppm
- Mercury (Hg):Max. 10 ppb
- Holmium (Ho):Max. 0.01 ppm
- Indium (In):Max. 0.005 ppm
- Iridium (Ir):Max. 0.5 ppm
- Potassium (K):Max. 0.5 ppm
- Lanthanum (La):Max. 0.01 ppm
- Lithium (Li):Max. 0.1 ppm
- Lutetium (Lu):Max. 0.01 ppm
- Magnesium (Mg):Max. 0.05 ppm
- Manganese (Mn):Max. 0.01 ppm
- Molybdenum (Mo):Max. 0.01 ppm
- Sodium (Na):Max. 0.2 ppm
- Neodymium (nd):Max. 0.01 ppm
- Nickel (Ni):Max. 0.01 ppm
- Lead (Pb):Max. 0.01 ppm
- Palladium (Pd):Max. 0.5 ppm
- Praseodymium (Pr):Max. 0.01 ppm
- Platinum (Pt):Max. 0.5 ppm
- Rubidium (Rb):Max. 0.1 ppm
- Rhodium (Rh):Max. 0.5 ppm
- Ruthenium (Ru):Max. 0.5 ppm
- Antimony (Sb):Max. 0.05 ppm
- Scandium (Sc):Max. 0.01 ppm
- Samarium (Sm):Max. 0.01 ppm
- Strontium (Sr):Max. 0.05 ppm
- Terbium (Tb):Max. 0.01 ppm
- Thorium (Th):Max. 0.01 ppm
- Titanium (Ti):Max. 0.01 ppm
- Thulium (Tm):Max. 0.01 ppm
- Uran (U):Max. 0.01 ppm
- Vanadium (V):Max. 0.01 ppm
- Yttrium (Y):Max. 0.01 ppm
- Ytterbium (Yb):Max. 0.01 ppm
- Zinc (Zn):Max. 0.1 ppm
- Total S (as SO₄):Max. 10 ppm
- Bromide, chloride (as Cl):Max. 100 ppm
- Phosphate (PO₄):Max. 10 ppm
- Iodate (IO₃):Max. 2 ppm
- Total N:Max. 10 ppm
Specification Test Results
About this item
:
:259.81 g/mol
Merck Index:12,02056
Boiling Pt:1280 °C (1013 hPa)
Melting Pt:621 °C
Density:3,21 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-145-2
:GHS08
CAS Number:7789-17-5
Formula:CsI
MDL Number:MFCD00010962