Order Entry
Ireland
ContactUsLinkComponent
Copper(II) sulphate pentahydrate 20% (w/v) in aqueous solution
 
 
Copper(II) sulphate pentahydrate 20% (w/v) in aqueous solution
  VWRS5998.9020
 :  
 
 
Copper(II) sulphate pentahydrate 20% (w/v) in aqueous solution
  VWRS5998.9020
 :  

 

  • Pack type:
    Plastic container
  • Pk:
    20 L
  • Seq:
    5302

 

 

 :
 :249.69 g/mol
 :Ambient
 :231-847-6
 :3082
 :9,III
 :GHS07
 :GHS09
 :7758-99-8
 :CuSO₄·5H₂O
 :MFCD00149681