(4-Bromophenyl)acetic acid
Jump to:
About this item
Synonyms: 4-Bromophenylacetic acid, 2-(4-bromophenyl)acetic acid
- CAS Number:1878-68-8
- Formula:(C₆H₄Br)CH₂COOH
- MDL Number:MFCD00004342
- Molecular Weight:215.05 g/mol
- Boiling Pt:326 °C (1013 hPa)
- Melting Pt:94…98 °C
- EINECS:217-523-7
0 Options Available Below
Product Details & Documents
Synonyms: 4-Bromophenylacetic acid, 2-(4-bromophenyl)acetic acid
- CAS Number:1878-68-8
- Formula:(C₆H₄Br)CH₂COOH
- MDL Number:MFCD00004342
- Molecular Weight:215.05 g/mol
- Boiling Pt:326 °C (1013 hPa)
- Melting Pt:94…98 °C
- EINECS:217-523-7



